Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-HOP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SRA-1500-F ADI-SRA-1500-D
200062-852EA 577.7 USD InStock
200062-852 200062-850
Anti-HOP Mouse Monoclonal Antibody
Antibodies
Hsp70-Hsp90 Organizing Protein (HOP, p60) is an ~60kDa protein that is a critical intermediate component for the efficient maturation of steroid receptor complexes, serving to recruit Hsp90 to Hsp70-containing complexes. HOP contains three tetratricopeptide repeat (TPR) domains, TPR1, TPR2a, and TPR2b.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, mink, pig, rabbit, sheep and Xenopus HOP. Detects a band of ~60kDa by Western blot.

Type: Primary
Antigen: HOP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty