Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-VAMP2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-SV014-E VAM-SV014-C
200063-298EA 406 USD InStock
200063-298 200062-918
Anti-VAMP2 Mouse Monoclonal Antibody
Antibodies
Vesicle-associated membrane protein (VAMP), also known as synaptobrevin, is an 18 kDa integral membrane protein localized to the cytoplasmic surface of synaptic vesicles. Two VAMP homologs, VAMP1 and VAMP2, are differentially expressed in the nervous system. VAMP2 is found in neurons associated with autonomic, sensory and integrative functions, and in non-neuronal Langerhans islets and glomerular cells. VAMP is a component of a 20S complex implicated in vesicle docking and fusion, the formation of which involves the interaction of VAMP with syntaxin through an ATP-dependent reaction modulated by synaptotagmin, SNAP-25, alpha-SNAP, and NSF.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, hamster, pig and Xenopus VAMP2. Detects a band of ~18kDa by Western blot.

Type: Primary
Antigen: VAMP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty