Bovine;Human;Mouse;Rat Orexin A
Supplier: SAFE AND SURE NEDERLAND B.V.
164-001-C100 164-001-M001
89143-642EA 204.92 USD InStock
89143-642 89143-646
Bovine;Human;Mouse;Rat Orexin A
Proteins and Peptides
Food intake modulator
Hypothalamic neuropeptide that stimulates food intake in rats with impressive potency. Binds specifically to the OX1 receptor.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty