Protein Kinase C (19-31)
Supplier: SAFE AND SURE NEDERLAND B.V.
165-009-M001 165-009-M005 BML-P206-0001
89143-682EA 64.4 USD InStock
89143-682 89143-684 89161-076
Protein Kinase C (19-31)
Proteins and Peptides
Inhibitor of protein kinase C (PKC).
Shorter and more potent inhibitor of PKC (IC50=100 nM) than Protein Kinase C (19-36), Pseudosubstrate Inhibitor (BML-P150).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty