E-64
Supplier: SAFE AND SURE NEDERLAND B.V.
260-007-M005 260-007-M025
89146-570EA 120.99 USD InStock
89146-570 89146-572
E-64
Proteins and Peptides
Protease inhibitor
Irreversible inhibitor of cysteine proteinases like papain, calpain, cathepsin B, L and H. Acts by forming a thioether bond with the thiol of the active cysteine. Does not inhibit serine proteinases. Inhibits degradation of autophagic cargo in autophagolysosomes.
Synthetic.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty