AC-IETD-AFC
Supplier: SAFE AND SURE NEDERLAND B.V.
260-110-M005 260-110-M010
89146-856EA 257.24 USD InStock
89146-856 89146-858
AC-IETD-AFC
Proteins and Peptides
Caspase and granzyme substrate
Fluorogenic substrate for caspase-8/caspase-3 processing enzyme and granzyme B. Also a substrate for caspase-10. The IETD sequence is based on caspase-3 proenzyme cleavage site.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty