Pifithrin-α (cyclic) hydrobromide ≥95% (by HPLC)
Supplier: SAFE AND SURE NEDERLAND B.V.
270-287-M010 270-287-M050
89147-568EA 240.89 USD InStock
89147-568 76001-972
Pifithrin-α (cyclic) hydrobromide ≥95% (by HPLC)
Pifithrin-α (cyclic) hydrobromide
p53 inhibitor
Stable analog of pifithrin-alpha (Prod. No. BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor.
CAS Number: 60477-34-1
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty