Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PI103 ≥97% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

270-460-M001 270-460-M005
89147-964EA 64.31 USD InStock
89147-964 89147-966
PI103 ≥97% (by HPLC)
PI103
Potent, cell permeable and ATP-competitive inhibitor of DNA-PK, phosphoinositide 3-kinase (PI(3)K), the rapamycin-sensitive (mTORC1) and rapamycin-insensitive (mTORC2) complexes of the protein kinase mTOR. Exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10µM. Effectively blocks cell proliferation in glioma cell lines both in vitro and in vivo.

  • Inhibitor of DNA-PK, PI3 kinase and mTOR

Off-white solid.

Soluble in DMSO (5mg/ml; warm).


Formula: C₁₉H₁₆N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD11983145
CAS Number: 371935-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty