Microcystin-RR
Supplier: SAFE AND SURE NEDERLAND B.V.
350-043-C050 350-043-C100 350-043-C250 350-043-C500 350-043-M001
89148-966EA 83.93 USD InStock
89148-966 89148-968 89148-970 89148-972 89148-974
Microcystin-RR
Proteins and Peptides
PP2A inhibitor
Arg-Arg analog of microcystin-LR (Prod. No. ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A).
Isolated from Microcystis aeruginosa.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty