Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rotenone ≥98% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

350-360-G001 350-360-G005
89149-586EA 88.29 USD InStock
89149-586 89149-588
Rotenone ≥98% (by HPLC)
Rotenone
Insecticide. Inhibitor of mitochondrial electron transport. Specifically inhibits NAD-linked substrate oxidation at the oxygen side of NADH dehydrogenase (EC50=10pmol/mg). Inhibits mammalian cell proliferation by inhibiting microtubule assembly through tubulin binding. Can inhibit mitochondrial swelling and cytochrome c release induced by N-methyl-4-phenylpyridinium.

Synthetic. Originally isolated from Lonchocarpus sp. or Derris sp.


Formula: C₂₃H₂₂O₆
MDL Number: MFCD09025614
CAS Number: 83-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty