Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Reveromycin A (from Streptomyces sp. MST-AS5344) ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

380-216-MC01 380-216-MC25
89150-216EA 285.58 USD InStock
89150-216 89150-218
Reveromycin A (from Streptomyces sp. MST-AS5344) ≥99% (by HPLC)
Reveromycin A
Antibiotic

Dominant analog of the reveromycin complex. Inhibitor of the mitogenic activity of epidermal growth factor (EGF). G1 phase cell cycle inhibitor, selectively inhibiting isoleucyl-tRNA synthetase. Displays antiproliferative as well as potent antifungal activity. Induces apoptosis in osteoclasts thus inhibiting bone resorption.

White to tan solid

Soluble in ethyl acetate, 100% ethanol, methanol, dimethyl formamide or DMSO

Envase: Shipping temperature: Ambient


Formula: C₃₆H₅₂O₁₁
MW: 660.8 g/mol
Storage Temperature: Freezer
CAS Number: 134615-37-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty