Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

UCN-01 (from Streptomyces sp. MST-AS5345) ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

380-222-M001 380-222-MC25
89150-230EA 559 USD InStock
89150-230 89150-232
UCN-01 (from Streptomyces sp. MST-AS5345) ≥99% (by HPLC)
UCN-01
Inhibitor of protein kinase C (PKC) and cyclin-dependent kinases (CDKs) resulting in accumulation of cells in the G1 phase and induction of apoptosis. Enhances the cytotoxicity of other anticancer drugs, such as DNA-damaging agents and anti-metabolite drugs, through putative abrogation of G2 and/or S phase accumulation induced by these agents.

  • PKC and CDK inhibitor

White film.

Soluble in methanol, 100% ethanol, dimethyl formamide or DMSO.


Formula: C₂₈H₂₆N₄O₄
MW: 482.5 g/mol
Storage Temperature: Freezer
CAS Number: 112953-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty