Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GW501516 ≥98% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

420-032-M001 420-032-M005
89150-762EA 105.73 USD InStock
89150-762 89150-764
GW501516 ≥98% (by HPLC)
GW501516
PPARδ agonist.

  • Formulation: White to off-white solid.

GW501516 is a specific agonist for PPARδ (EC₅₀ = 1.1 nM) with a 1000-fold selectivity over the other human subtypes.

Soluble in DMSO (50 mg/mL), dimethyl formamide (25 mg/mL), 100% ethanol (12 mg/mL) or methanol (50 mg/mL); slightly soluble in water.


Formula: C₂₁H₁₈F₃NO₃S₂
MW: 453.494 g/mol
Storage Temperature: Refrigerator
CAS Number: 317318-70-0
Order Now
Specification Test Results
Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR and ¹³C-NMR

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty