Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcimycin (mixed Mg/CA salt) ≥98% (by TLC)

Danger

450-002-M005 450-002-M010
89151-370EA 118 USD InStock
89151-370 89151-372
Calcimycin (mixed Mg/CA salt) ≥98% (by TLC)
Calcimycin (mixed Mg/CA salt)
Salt form of mixed calcium-magnesium. Used in cell activation experiments when calcium dose-response data are not required.

  • Formulation: White to off-white crystalline solid.
  • Isolated from Streptomyces chartreusensis.
  • Semisynthetic

Soluble in methylene chloride, chloroform, acetone, ethyl acetate, or diethyl ketone; also soluble in DMSO, 100% ethanol, and ethanol. Solubilize in DMSO or ethanol prior to preparing aqueous solutions of very low concentrations.


Formula: (C₂₉H₃₆N₃O₆)₂·Mg(C₂₉H₃₆N₃O₆)₂·Ca
Storage Temperature: Refrigerator
CAS Number: 52665-69-7
Merck Index: 13,01639
Order Now
Specification Test Results
Purity (TLC) ≥98%
Source Isolated from Streptomyces chartreusensis

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty