Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Loxoribine ≥97% (by HPLC and UV method)

Supplier: SAFE AND SURE NEDERLAND B.V.

480-097-M025 480-097-M100
89151-598EA 203.83 USD InStock
89151-598 89151-600
Loxoribine ≥97% (by HPLC and UV method)
Loxoribine
Selective ligand for toll-like receptor 7 (TLR7) via IFN.

  • Formulation: White to off-white solid.

Activates natural killer cells and primes cytolytic precursor cells for activation by IL-2.

Soluble in DMSO (30 mg/mL), water (10 mg/mL), water:acetonitrile (1:1) (20 mg/mL) or water:methanol (1:1) (10 mg/mL).


Formula: C₁₃H₁₇N₅O₆
Storage Temperature: Freezer
MDL Number: MFCD00867591
CAS Number: 121288-39-9
Order Now
Specification Test Results
Purity (HPLC (UV)) ≥97%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty