Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gambogic acid ≥95% (by TLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-AP305-0005 BML-AP305-0025
89156-586EA 118.81 USD InStock
89156-586 89156-588
Gambogic acid ≥95% (by TLC)
Gambogic acid
Gambogic Acid is a natural product isolated from Garcinia Hurburyi. It induces apoptosis in several tumor cell lines including T47D breast cancer cells and MGC-803 human gastric carcinoma cells but not in normal cells. Apoptosis induction proceeds via upregulation of bax and inhibition of bcl-2 expression.The target of gambogic acid has been identified as the transferrin receptor.

  • Bcl-2 inhibitor, Bax activator

Yellow-orange solid

Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml)


Formula: C₃₈H₄₄O₈
Storage Temperature: Freezer
MDL Number: MFCD16878985
CAS Number: 2752-65-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty