Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-EI178-0250
89157-720EA 121 USD InStock
89157-720 76002-074
N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (by HPLC)
N-(4-n-Pentylcinnamoyl)anthranilic acid
ACA is a close structural analog of the PLA2 inhibitor, ONO-RS-082 and possesses similar but less potent inhibitory activity. Inhibits TRP channels independently of PLA2, including TRPM2, TRPC6 and TRPM8. It is a useful tool for studying the physiological role of TRP channels.

  • Inhibitor of PLA2 and TRP channels

White solid.

Soluble in DMSO (50 mg/ml) or ethanol (50 mg/ml @ 60°C).


Formula: C₂₁H₂₃NO₃
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00211365
CAS Number: 110683-10-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty