Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxy-2-naphthalenylmethylphosphonic acid ≥98% (by TLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-EI247-0005 BML-EI247-0025
89157-862EA 99.19 USD InStock
89157-862 89157-864
Hydroxy-2-naphthalenylmethylphosphonic acid ≥98% (by TLC)
Hydroxy-2-naphthalenylmethylphosphonic acid
HNMPA is a membrane impermeable inhibitor of insulin receptor tyrosine kinase (IC50 = 100µM); in rat adipocytes (IC50 = 10µM). Inhibits serine and tyrosine autophosphorylation by the human insulin receptor.

  • Insulin receptor inhibitor

White solid.

Soluble in 100% ethanol (25mg/ml), DMSO (25ml/ml) or 5% Sodium bicarbonate (25mg/ml).


Formula: C₁₁H₁₁O₄P
MW: 238.2 g/mol
Melting Pt: ≥250 °C
Storage Temperature: Freezer
CAS Number: 132541-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty