Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CCG-1423 ≥97% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-EI394-0010 BML-EI394-0050
89158-262EA 99.19 USD InStock
89158-262 89158-264
CCG-1423 ≥97% (by HPLC)
CCG-1423
Cell permeable inhibitor of RhoA- and RhoC-mediated signaling. Inhibits LPA-induced DNA synthesis in PC-3 prostate cancer cell lines (<1µM) and inhibits the growth of RhoC-overexpressing melanoma cell lines (A375M2 and SK-Mel-147) at nanomolar concentrations.

  • Rho inhibitor

White solid.

Soluble in DMSO (10mg/ml) or 100% ethanol (9mg/ml).


Formula: C18H13ClF6N2O3
Storage Temperature: Ambient
MDL Number: MFCD01566719
CAS Number: 285986-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty