Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ENMD-1068

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-N110-0005 BML-N110-0025
89160-582EA 139.52 USD InStock
89160-582 89160-584
ENMD-1068
ENMD-1068
Proteinase-activated receptor ligand

Selective PAR-2 antagonist. Dose dependently inhibits calcium signaling in Lewis lung carcinoma cells stimulated with a soluble peptide PAR-2 agonist or trypsin, but not with a soluble PAR-1 agonist. Does not inhibit platelet aggregation stimulated by thrombin suggesting that it does not inhibit PAR-3 or PAR-4 signaling. Reduces TNF-α and IL-1β production from rheumatoid arthritis synovial primary cell culture and reduces joint swelling in a carageenan/kaolin-injected mouse model.


Formula: C₁₅H₂₉N₃O₂·HCl
CAS Number: 789488-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty