Z-Leu-Leu-Cho
Supplier: SAFE AND SURE NEDERLAND B.V.
BML-PI116-0005 BML-PI116-0025
89161-608EA 107.91 USD InStock
89161-608 89161-610
Z-Leu-Leu-Cho
Proteins and Peptides
Calpain inhibitor
Selective inhibitor of calpain, 100-fold selective for calpain over proteasome.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty