Aaf-Cmk
Supplier: SAFE AND SURE NEDERLAND B.V.
BML-PI123-0005 BML-PI123-0025
89161-620EA 98.1 USD InStock
89161-620 89161-622
Aaf-Cmk
Proteins and Peptides
Tripeptidyl peptidase inhibitor
A serine protease inhibitor which irreversibly inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions, and reversibly inhibits TPPI. Shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty