Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Proteasome 19S Rpn10/S5A Subunit Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW9250-0025 PW9250-0100
89162-308EA 189.66 USD InStock
89162-308 89162-310
Anti-Proteasome 19S Rpn10/S5A Subunit Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human proteasome 26S (19S regulator complex) (Rpn10/S5a subunit).

Type: Primary
Antigen: PROTEASOME 19S RPN10/S5A SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty