Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Picotamide ≥98%

Supplier: SAFE AND SURE NEDERLAND B.V.

BML-RA104-0010 BML-RA104-0050
89162-476EA 118.81 USD InStock
89162-476 89162-478
Picotamide ≥98%
Picotamide
Thromboxane A2 antagonist and TXA2 synthase inhibitor

A dual activity thromboxane A2 antagonist and TXA2 synthase inhibitor. Clinically useful antiplatelet agent.


Formula: C₂₁H₂₀N₄O₃
MDL Number: MFCD00866792
CAS Number: 32828-81-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty