Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-RELM-beta Mouse Monoclonal Antibody [clone: HRB 149]

Supplier: Adipogen
AG-20A-0012-C050 AG-20A-0012-C100
102513-952EA 138.46 USD InStock
102513-952 102513-954
Anti-RELM-beta Mouse Monoclonal Antibody [clone: HRB 149]
Antibodies
RELM-beta is a 19kDa disulfide-linked homodimeric protein expressed in the epithelium of the colon and small bowel. RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin.

Type:
Antigen: RELM-beta
Clonality: Monoclonal
Clone: HRB 149
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty