Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Vaspin Mouse Monoclonal Antibody [clone: VP63]

Supplier: Adipogen
AG-20A-0045-C050 AG-20A-0045-C100
102514-090EA 283.85 USD InStock
102514-090 102514-092
Anti-Vaspin Mouse Monoclonal Antibody [clone: VP63]
Antibodies
Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new_x000D_ therapeutic target of metabolic syndrome.

Type:
Antigen: Vaspin
Clonality: Monoclonal
Clone: VP63
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty