Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ANGPTL4 Mouse Monoclonal Antibody [clone: Kairos4-153AD]

Supplier: Adipogen
AG-20A-0046-C050 AG-20A-0046-C100
102514-094EA 138.46 USD InStock
102514-094 102514-096
Anti-ANGPTL4 Mouse Monoclonal Antibody [clone: Kairos4-153AD]
Antibodies
ANGPTL4 mainly expressed in endothelial cells (hypoxia-induced). Regulates angiogenesis and modulates tumorgenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage.

Type:
Antigen: ANGPTL4
Clonality: Monoclonal
Clone: Kairos4-153AD
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty