Human Recombinant FGF23 (from HEK293 cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40A-0109-C010 AG-40A-0109-C050
102515-080EA 235.38 USD InStock
102515-080 102515-082
Human Recombinant FGF23 (from HEK293 cells)
Proteins and Peptides
FGF-23 (Fibroblast growth factor 23) is a regulator of phosphate homeostasis. It upregulates EGR1 expression in the presence of KLBy. Acts directly on the parathyroid to decrease PTH secretion. Regulates the vitamin-D metabolism. Negatively regulates osteoblast differentiation and matrix mineralization. Defects in FGF-23 are the cause of autosomal dominant hypophosphataemic rickets (ADHR) and of hyperphosphatemic familial tumoral calcinosis (HFTC).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty