Human Recombinant CALRETICULIN (from E. coli)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40A-0132-C010 AG-40A-0132-C050
102515-160EA 276.92 USD InStock
102515-160 102515-162
Human Recombinant CALRETICULIN (from E. coli)
Proteins and Peptides
Calreticulin is involved in regulation of intracellular Ca2+ homoeostasis and ER Ca2+ capacity. It constitutes together with calnexin and ERp57 the ‘calreticulin/calnexin cycle’ that is responsible for folding and quality control of newly synthesized glycoproteins. Calreticulin has been implicated to play a role in many biological systems, including functions inside and outside the ER, indicating that the protein is a multi-process molecule. Recently, Calreticulin was shown to enhance the merger of macrophages and tumor cells, increasing phagocytosis.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty