Human Recombinant Il-37 (from E. coli)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40A-0190-C010 AG-40A-0190-C050
102515-372EA 415.38 USD InStock
102515-372 102515-374
Human Recombinant Il-37 (from E. coli)
Proteins and Peptides
IL-37 (IL-1F7; IL-1H4) is an IL-1 family member that is expressed only in certain types of human organs and cells such as heart, thymus, testis, kidney, mononuclear cells (PBMCs) and dendritic cells. IL-37 is an inhibitor of innate immunity with anti-inflammatory properties. It binds to the interleukin-18 receptor (IL-18R) and its co-receptor SIGIRR. IL-37 is secreted as a full-length and as a processed form starting from amino acid Val46. It plays an important role as a link between innate and adaptive immunity and acts as an inhibitor of autoimmune diseases and tumor growth. IL-37 also inhibited Lipopolysaccharide (LPS)-induced immunological reaction and LPS-induced osteoclast formation and bone resorption.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty