Human Recombinant DR6 (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0011-C050 AG-40B-0011-3050
102515-426EA 193.85 USD InStock
102515-426 102515-428
Human Recombinant DR6 (from HEK293 Cells)
Proteins and Peptides
DR6 is an orphan TNF receptor superfamily member belonging to a subgroup of receptors called death receptors. Expressed ubiquitously with high expression in lymphoid organs, heart, brain, and pancreas. Broadly expressed by developing neurons where it functions as pro-apoptotic factor. Recently, it has been reported that interaction with the beta-amyloid precursor protein (APP) activates a widespread caspase-dependent self-destruction program dependent on caspase-6.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty