Human Recombinant OX40 (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0014-C050 AG-40B-0014-3050
102515-430EA 249.23 USD InStock
102515-430 102515-432
Human Recombinant OX40 (from HEK293 Cells)
Proteins and Peptides
OX40 binds to receptors on T-cells, preventing them from dying and subsequently increasing cytokine production. OX40 has a critical role in the maintenance of an immune response. OX40:Fc was shown to prevent OX40 from reaching the T-cell receptors, thus reducing the T-cell response. Experiments in mice have demonstrated that OX40:Fc can reduce the symptoms associated with the cytokine storm (an immune overreaction) while allowing the immune system to fight off the virus successfully.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty