Human Recombinant RANK (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0018-C050 AG-40B-0018-3050
102515-442EA 276.92 USD InStock
102515-442 102515-444
Human Recombinant RANK (from HEK293 Cells)
Proteins and Peptides
RANK (TRANCE-R; CD265) is the receptor for RANKL/TRANCE/OPGL. It is essential for RANKL-mediated osteoclastogenesis and is involved in the regulation of interactions between T cells and dendritic cells.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty