Human Recombinant GITR (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0028-C050 AG-40B-0028-3050
102515-466EA 443.08 USD InStock
102515-466 102515-468
Human Recombinant GITR (from HEK293 Cells)
Proteins and Peptides
GITR (Glucocorticoid-induced TNF receptor) is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T cell receptor-mediated cell death. GITR serves as a negative regulator for NK cell activation. It mediates NF-kappaB activation via the TRAF2/NIK pathway.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty