Human Recombinant UNC5B (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0037-C050 AG-40B-0037-3050
102515-484EA 332.31 USD InStock
102515-484 102515-486
Human Recombinant UNC5B (from HEK293 Cells)
Proteins and Peptides
UNC5B is a receptor for the netrin family of guidance molecules. It was identified as a mediator of the repulsive response in neurons and it is the only known netrin receptor with prominent endothelial expression. It has an important role in angiogenesis signaling.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty