Human Recombinant TRAILR2 (from HEK293 Cells)
Supplier: Van Der Voort Lucht en Milieu Amste
AG-40B-0071-C050 AG-40B-0071-3050
102515-534EA 249.23 USD InStock
102515-534 102515-536
Human Recombinant TRAILR2 (from HEK293 Cells)
Proteins and Peptides
TRAIL-R2 (DR5; CD262) is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty