Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferulenol ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0011-M001 AG-CN2-0011-M005 AG-CN2-0011-M010
102515-700EA 55.38 USD InStock
102515-700 102515-702 102515-704
Ferulenol ≥98% (by HPLC)
Ferulenol
Prenylated 4-hydroxycoumarin. Anti-tumor compound. Cytotoxic. Stimulator of tubulin polymerization in vitro. Inhibitor of colchicine binding to tubulin. Antitubercular antibiotic with potent antibacterial activity. Anti-coagulant, pro-haemorrhagic compound with higher activity than warfarin. Shows hepatocyte toxicity. Disrupts mitochondrial membrane potential.


Formula: C₂₄H₃₀O₃
Storage Temperature: Freezer
CAS Number: 6805-34-1
Order Now
Specification Test Results
Source/Host Isolated from Ferula communis.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in 100% ethanol, methanol or DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty