Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

K-252a (from Nonomuraea longicatena chiba) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0019-C100 AG-CN2-0019-M001
102515-726EA 83.08 USD InStock
102515-726 102515-728
K-252a (from Nonomuraea longicatena chiba) ≥98% (by HPLC)
K-252a
K-252a is an antibiotic, PKC, PKA and PKG inhibitor.

  • Highly potent cell permeable CaM kinase and phosphorylase inhibitor
  • Apoptosis inducer
  • Potential drug for psoriasis treatment

Antibiotic. PKC, PKA and PKG inhibitor. Shows neurotropic activity by inducing neurite outgrowth. Tyrosine protein kinase inhibitor of the TRK family. Antiproliferative. Cell cycle arrest inducer.

White to off-white powder

Soluble in DMSO, methanol, methylene chloride, ethyl acetate or acetone. Insoluble in water.


Formula: C₂₇H₂₁N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00161522
CAS Number: 97161-97-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty