Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SIN-1 chloride ≥98% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CR1-0027-M010 AG-CR1-0027-M050
102515-974EA 41.54 USD InStock
102515-974 102515-976
SIN-1 chloride ≥98% (by NMR)
SIN-1 chloride
Nitric oxide (NO) donor. Generates both superoxide anion and nitric oxide that spontaneously form peroxynitrite. Platelet aggregation inhibitor. Guanylyl cyclase activator. Cytotoxic. Positive ionotropic. Vasodilator.


Formula: C₆H₁₁ClN₄O₂
Storage Temperature: Freezer
CAS Number: 16142-27-1
Order Now
Specification Test Results
Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in 100% ethanol, water or DMSO ( 5 mg/ml).

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty