Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SB-366791 ≥99%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CR1-0034-M005 AG-CR1-0034-M025
102516-012EA 76.15 USD InStock
102516-012 102516-014
SB-366791 ≥99%
SB-366791
Potent and selective TRPV1 antagonist. Glutamatergic synaptic transmission inhibitor.


Formula: C₁₆H₁₄ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD01033818
CAS Number: 472981-92-3
Order Now
Specification Test Results
Purity 99% (NMR)
Appearance/Color White to beige crystalline solid.
Solubility Soluble in 100% ethanol (warm) or DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty