Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclic pifithrin-alpha-hydrobromide ≥98% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CR1-0052-M005 AG-CR1-0052-M010 AG-CR1-0052-M050
102516-078EA 69.23 USD InStock
102516-078 102516-080 102516-082
Cyclic pifithrin-alpha-hydrobromide ≥98% (by NMR)
Cyclic pifithrin-alpha-hydrobromide
Stable analog of pifithrin-alpha. p53 inhibitor.


Formula: C₁₆H₁₆N₂S
Storage Temperature: Freezer
MDL Number: MFCD02683960
CAS Number: 511296-88-1
Order Now
Specification Test Results
Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty