Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H₂S Donor 5a ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CR1-3510-M005 AG-CR1-3510-M025
102516-244EA 110.77 USD InStock
102516-244 102516-246
H₂S Donor 5a ≥98%
H2S Donor 5a
Cysteine-activated hydrogen sulfide (H2S) donor. Regulates the rate of H2S generation.


Formula: C₁₄H₁₁NO₂S
MW: 257.3 g/mol
Storage Temperature: Freezer
CAS Number: 134861-13-5
Order Now
Specification Test Results
Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty