Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Swinholide A ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0035-C010 AG-CN2-0035-C050
102515-798EA 135.69 USD InStock
102515-798 102515-800
Swinholide A ≥97%
Swinholide A
Cell permeable dimeric dilactone macrolide. Cytotoxic compound. Specific actin filament (F-actin) inhibitor. Disrupts the actin cytoskeleton by sequestering actin dimers and rapidly severing actin filaments (F-actin). Depolymerizes actin filaments (F-actin). Antifungal. Apoptosis inducer. Alterates the actin cytoskeleton of GH4 cells and inhibits insulin-increased prolactin gene transcription.


Formula: C₇₈H₁₃₂O₂₀
MW: 1389.9 g/mol
Storage Temperature: Freezer
CAS Number: 95927-67-6
Order Now
Specification Test Results
Source/Host Isolated from marine sponge Theonella swinhoei.
Purity 97% (HPLC)
Appearance/Color Colorless oil.
Solubility Soluble in ethanol, methanol, DMSO, acetone or ethyl acetate.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty