Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CR1-3566-M010 AG-CR1-3566-M050 AG-CR1-3566-M250
102516-298EA 27.69 USD InStock
102516-298 102516-300 102516-302
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥98%
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid
Potent peroxisome proliferator-activated receptor (PPARalpha) activator. Activates also PPARgamma but not PPARdelta. Potent anti-hypercholesterolemic agent. Hypolipidemic compound. Lipogenesis inducer. Tumor promoter. Causes increased cell proliferation and decreased apoptosis. Anti-inflammatory. Inhibits NF-kappaB transcriptional activity and decreases the inflammatory response by reducing the production of inflammatory cytokines (TNF-alpha, IL1beta). Reduces oxidative stress. Increases fatty acid oxidation. Directly affects insulin signaling. Increases glucose uptake. Reviews.


Formula: C₁₄H₁₄ClN₃O₂S
MW: 323.80 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00191335
CAS Number: 50892-23-4
Order Now
Specification Test Results
Purity 98%
Appearance/Color Off-white solid.
Solubility Soluble in ethanol, DMSO, dimethyl formamide or PBS (pH 7.2).

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty