Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imiquimod ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CR1-3569-M100 AG-CR1-3569-M250
102516-314EA 55.38 USD InStock
102516-314 102516-316
Imiquimod ≥98%
Imiquimod
Immune response modulator. Anti-inflammatory. TLR7 agonist. Stimulates proinflammatory cytokine production interferon-alpha (IFN-alpha), interleukin-6 (IL-6) and tumor necrosis factor-alpha (TNF-alpha) and activates NF-kappaB. Caspase-3 activator that directly induces procaspase-3 cleavage. Anti-cancer compound. Apoptosis inducer. Antiproliferative, independent of immune system activation or function. Antiviral and anti-angiogenic. Approved treatment for external genital warts caused by human papillomavirus infection.


Formula: C₁₄H₁₆N₄
MW: 240.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866946
CAS Number: 99011-02-6
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Purity 98%
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO or dimethylformamide. Insoluble in ethanol. Slightly soluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty