Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Etoposide ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CR1-3572-M025 AG-CR1-3572-M100 AG-CR1-3572-5100
102516-330EA 34.62 USD InStock
102516-330 102516-332 102516-334
Etoposide ≥98%
Etoposide
Potent anti-cancer compound. Induces apoptosis in normal and tumor cell lines. DNA Topoisomerase II activity inhibitor. Increases Topo II-mediated DNA breakage primarily by inhibiting the ability of the enzyme to religate cleaved nucleic acid molecules. Does not lead to immediate block of DNS synthesis, induces a progressive inhibition of DNA replication. p53 activator. Blocks the cell cycle between the end of the S phase and the early G2 phase. Oncoprotein Mdm2 synthesis inhibitor. Apoptosis inducer through the cytochrome c/Apaf-1/caspase-9 pathway and the Fas-mediated death signaling pathway. Cell cycle checkpoint activator. Affects gene expression at different levels (chromatin remodeling, transcription and alternative splicing). Chemotherapeutic compound used in cancers. Used in conditioning regimen prior to a bone marrow or blood stem cell transplantation. Highly effective in mobilizing stem cells


Formula: C₂₉H₃₂O₁₃
MW: 588.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00869325
CAS Number: 33419-42-0
Order Now
Specification Test Results
Source/Host Semisynthetic derivative of podophyllotoxin.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO, ethanol, methanol or dimethylformamide.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty