Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Paclitaxel ≥99%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CN2-0045-M001 AG-CN2-0045-M005 AG-CN2-0045-M025 AG-CN2-0045-M100
102515-830EA 30.46 USD InStock
102515-830 102515-832 102515-834 102515-836
Paclitaxel ≥99%
Paclitaxel
Anticancer compound. Chemotherapeutic used in patients with cancer and advanced forms of Kaposi's sarcoma. Microtubule assembly stabilizer. Reversibly binds to polymerized tubulin. Anti-mitotic. Mitotic spindle assembly, chromosome segregation and cell division inhibitor. Induces cell cycle arrest at the G2/M phase. Apoptosis inducer through aberrant activation of cyclin-dependent kinases (CKDs) and the c-Jun N-terminal kinase/stress activated protein kinase (JNK/SAPK). Immunosuppressor. Immunostimulant. Antiproliferative agent for the prevention of restenosis. TRAIL sensitizer


Formula: C₄₇H₅₁NO₁₄
Storage Temperature: Freezer
MDL Number: MFCD00869953
CAS Number: 33069-62-4
Order Now
Specification Test Results
Source/Host Isolated from the bark of the pacific yew tree (Taxus brevifolia).
Purity 99% (HPLC)
Appearance/Color White powder.
Solubility Soluble in DMSO, ethanol or acetonitrile.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty