Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Echinosporin (from Streptomyces species) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0006-M001 BVT-0006-M005
102513-446EA 180 USD InStock
102513-446 102513-448
Echinosporin (from Streptomyces species) ≥98% (by HPLC)
Echinosporin
Echinosporin is a cell cycle inhibitor at the G(2)/M phase.

  • Antibiotic
  • Apoptosis inducer

Antibiotic. Cell cycle inhibitor at the G(2)/M phase. Anti-tumor compound. Apoptosis inducer.

White to off-white solid

After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C. Stock solutions are stable for at least three months when stored at -20°C.

Soluble in water, methanol or dimethylformamide.


Formula: C₁₀H₉NO₅
MW: 223.2 g/mol
Storage Temperature: Freezer
CAS Number: 79127-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty