Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aspyrone ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0029-M001 BVT-0029-M005
102513-470EA 110.77 USD InStock
102513-470 102513-472
Aspyrone ≥98% (by HPLC)
Aspyrone
Aspyrone shows nematicidal, insecticidal, antibacterial and antifungal activity.

  • Antibiotic
  • Related to asperlactone and aspinonene
  • Anti-bacterial and anti-fungal activity

Antibiotic. Shows nematicidal, insecticidal, antibacterial and antifungal activity. Related to asperlactone and aspinonene.

White to off-white solid

After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.

Soluble in DMSO, acetone or methanol.


Formula: C₉H₁₂O₄
MW: 184.2 g/mol
Storage Temperature: Freezer
CAS Number: 17398-00-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty