Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CJ-21058 ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

BVT-0064-M001 BVT-0064-M005
102513-490EA 166.15 USD InStock
102513-490 102513-492
CJ-21058 ≥98%
CJ-21058
Antibiotic. SecA inhibitor. Antibacterial and antifungal. Cytotoxic. Methyl analog of equisetin.


Formula: C₂₃H₃₃NO₄
Storage Temperature: Freezer
CAS Number: 405072-57-3
Order Now
Specification Test Results
Source/Host Isolated from Phoma sp.
Purity 98% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in methylene chloride, DMSO or methanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty